missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Benzyladenine, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Inhibitor of respiratory kinase in plants. Increases post-harvest life of green vegetables6-Benzyladenine is a plant growth regulator used to stimulate growth in apples, melons, oranges, roses, pears, sweet cherries and white pine trees. It is also used to establish and screen the optimal rapid propagation technology of S. tonkinensis by orthogonal test.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Inhibitor of respiratory kinase in plants. Increases post-harvest life of green vegetables6-Benzyladenine is a plant growth regulator used to stimulate growth in apples, melons, oranges, roses, pears, sweet cherries and white pine trees. It is also used to establish and screen the optimal rapid propagation technology of S. tonkinensis by orthogonal test.
Solubility
Soluble in water, methanol and acetone. Slightly soluble in ethyl acetate and dichloromethane and toluene. Insoluble in n-hexane.
Notes
Incompatible with strong oxidizing agent.
Inhibitor of respiratory kinase in plants. Increases post-harvest life of green vegetables6-Benzyladenine is a plant growth regulator used to stimulate growth in apples, melons, oranges, roses, pears, sweet cherries and white pine trees. It is also used to establish and screen the optimal rapid propagation technology of S. tonkinensis by orthogonal test.
Solubility
Soluble in water, methanol and acetone. Slightly soluble in ethyl acetate and dichloromethane and toluene. Insoluble in n-hexane.
Notes
Incompatible with strong oxidizing agent.
Specifications
Specifications
Melting Point | 229°C to 233°C |
Assay Percent Range | 99% |
Quantity | 5 g |
Beilstein | 19406 |
Solubility Information | Soluble in water,methanol and acetone. Slightly soluble in ethyl acetate and dichloromethane and toluene. Insoluble in n-hexane. |
Formula Weight | 225.26 |
Percent Purity | 99% |
Chemical Name or Material | 6-Benzyladenine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'