missing translation for 'onlineSavingsMsg'
Learn More

6-(Benzyloxy)-2-naphthylboronic acid, 97%, Thermo Scientific™

Product Code. 10394672
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 152915-83-8
Molecular Formula C17H15BO3
Molecular Weight (g/mol) 278.114
MDL Number MFCD04115402
InChI Key ZVHAZMUBLCEMGG-UHFFFAOYSA-N
Synonym 6-benzyloxy-2-naphthylboronic acid, 6-benzyloxynaphthalene-2-boronic acid, 6-benzyloxy naphthalen-2-ylboronic acid, boronic acid,b-6-phenylmethoxy-2-naphthalenyl, 6-benzyloxy-2-naphthaleneboronic acid, 6-benzyloxy naphthalen-2-yl boronic acid, 6-phenylmethoxynaphthalen-2-yl boronic acid, acmc-209d8c, 6-benzyloxy-2-naphthylboronicacid
PubChem CID 2794628
IUPAC Name (6-phenylmethoxynaphthalen-2-yl)boronic acid
SMILES B(C1=CC2=C(C=C1)C=C(C=C2)OCC3=CC=CC=C3)(O)O

Specificatie

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 278.11
Percent Purity 97%
Chemical Name or Material 6-(Benzyloxy)-2-naphthylboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'