missing translation for 'onlineSavingsMsg'
Learn More

6-Boc-1-oxa-6-azaspiro[3.3]heptane, 96%, Thermo Scientific Chemicals

Product Code. 15493588
Click to view available options
Quantity:
250 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1223573-41-8
Molecular Formula C10H17NO3
Molecular Weight (g/mol) 199.25
MDL Number MFCD16877674
InChI Key QOXFRPSIASGKGE-UHFFFAOYSA-N
Synonym tert-butyl 1-oxa-6-azaspiro 3.3 heptane-6-carboxylate, 1-oxa-6-azaspiro 3.3 heptane-6-carboxylic acid tert-butyl ester, 6-boc-1-oxa-6-azaspiro 3.3 heptane, tert-butyl2,5-dioxa-8-aza-spiro 3,5 nonane-8-carboxylate, 2-aza-5-oxaspiro 3.3 heptane-2-carboxylic acid tert-butyl ester, 1-oxa-6-azaspiro 3.3 heptane-6-carboxylic acid,1,1-dimethylethyl ester, tert-butyl 1-oxa-6-azaspiro 3.3 heptane-6-carboxylate 95+%
PubChem CID 54758968
IUPAC Name tert-butyl 1-oxa-6-azaspiro[3.3]heptane-6-carboxylate
SMILES CC(C)(C)OC(=O)N1CC2(CCO2)C1

Specifications

Quantity 500 mg
Formula Weight 199.25
Percent Purity 96%
Chemical Name or Material 6-Boc-1-oxa-6-azaspiro[3.3]heptane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'