missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-1,4-dimethyl-1,2,3,4-tetrahydroquinoxaline, ≥97%, Thermo Scientific™

Product Code. 10694215
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 876728-35-7
Molecular Formula C10H13BrN2
Molecular Weight (g/mol) 241.13
MDL Number MFCD08271891
InChI Key BVWZHBZUOICVIU-UHFFFAOYSA-N
Synonym 6-bromo-1,4-dimethyl-1,2,3,4-tetrahydroquinoxaline, quinoxaline,6-bromo-1,2,3,4-tetrahydro-1,4-dimethyl, 6-bromanyl-1,4-dimethyl-2,3-dihydroquinoxaline
PubChem CID 18525730
IUPAC Name 6-bromo-1,4-dimethyl-2,3-dihydroquinoxaline
SMILES CN1CCN(C)C2=C1C=CC(Br)=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 241.13
Percent Purity 97%
Chemical Name or Material 6-Bromo-1,4-dimethyl-1,2,3,4-tetrahydroquinoxaline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'