missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-1-hexanol, 95%, Thermo Scientific Chemicals

Product Code. 10207550
Click to view available options
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4286-55-9
Molecular Formula C6H13BrO
Molecular Weight (g/mol) 181.07
MDL Number MFCD00002983
InChI Key FCMCSZXRVWDVAW-UHFFFAOYSA-N
Synonym 6-bromo-1-hexanol, 6-bromohexanol, 1-hexanol, 6-bromo, 6-bromo-hexan-1-ol, hexamethylene bromohydrin, 6-bromo-hexanol, 6-bromo-l-hexanol, 1-bromo-6-hexanol, 1-bromohexan-6-ol, 6-hydroxyhexyl bromide
PubChem CID 77970
IUPAC Name 6-bromohexan-1-ol
SMILES OCCCCCCBr

Specifications

CAS Min % 94.0
CAS Max % 100.0
Density 1.3200g/mL
Boiling Point 104.0°C to 106.0°C (5.0 mmHg)
Flash Point >110°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula Br(CH2)6OH
Refractive Index 1.4810 to 1.4840
Quantity 2.5 g
Beilstein 01,1704
Specific Gravity 1.32
Formula Weight 181.07
Percent Purity 95%
Chemical Name or Material 6-Bromo-1-hexanol
Show More Show Less
compliance-icons
Product Identifier
  • 6-Bromo-1-hexanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'