missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Bromo-1-indanone, 97%, Thermo Scientific Chemicals
£45.70 - £152.00
Chemical Identifiers
CAS | 14548-39-1 |
---|---|
Molecular Formula | C9H7BrO |
Molecular Weight (g/mol) | 211.058 |
MDL Number | MFCD02179286 |
InChI Key | SEQHEDQNODAFIU-UHFFFAOYSA-N |
Synonym | 6-bromo-1-indanone, 6-bromoindanone, 6-bromo-2,3-dihydro-1h-inden-1-one, 6-bromo-indan-1-one, 6-bromoindan-1-one, 1-indanone, 6-bromo, 1h-inden-1-one, 6-bromo-2,3-dihydro, 6-bromo indanone, 6-bromo-indanone |
PubChem CID | 139778 |
IUPAC Name | 6-bromo-2,3-dihydroinden-1-one |
SMILES | C1CC(=O)C2=C1C=CC(=C2)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15450438
|
Thermo Scientific Alfa Aesar
H54003.03 |
1 g |
£45.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15460438
|
Thermo Scientific Alfa Aesar
H54003.06 |
5 g |
£152.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
6-Bromo-1-indanone is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-Bromo-1-indanone is used as an organic chemical synthesis intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
14548-39-1 | |
211.058 | |
SEQHEDQNODAFIU-UHFFFAOYSA-N | |
139778 | |
C1CC(=O)C2=C1C=CC(=C2)Br |
C9H7BrO | |
MFCD02179286 | |
6-bromo-1-indanone, 6-bromoindanone, 6-bromo-2,3-dihydro-1h-inden-1-one, 6-bromo-indan-1-one, 6-bromoindan-1-one, 1-indanone, 6-bromo, 1h-inden-1-one, 6-bromo-2,3-dihydro, 6-bromo indanone, 6-bromo-indanone | |
6-bromo-2,3-dihydroinden-1-one |
Specifications
14548-39-1 | |
C9H7BrO | |
6-bromo-1-indanone, 6-bromoindanone, 6-bromo-2,3-dihydro-1h-inden-1-one, 6-bromo-indan-1-one, 6-bromoindan-1-one, 1-indanone, 6-bromo, 1h-inden-1-one, 6-bromo-2,3-dihydro, 6-bromo indanone, 6-bromo-indanone | |
C1CC(=O)C2=C1C=CC(=C2)Br | |
211.058 | |
211.06 | |
6-Bromo-1-indanone |
110°C | |
MFCD02179286 | |
SEQHEDQNODAFIU-UHFFFAOYSA-N | |
6-bromo-2,3-dihydroinden-1-one | |
139778 | |
97% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only