missing translation for 'onlineSavingsMsg'
Learn More
6-Bromo-3,4-dihydro-2H-1,4-benzothiazine, 97%, Thermo Scientific Chemicals Product Code.: 15437588

6-Bromo-3,4-dihydro-2H-1,4-benzothiazine, 97%, Thermo Scientific Chemicals

Product Code. 15437588
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15437588

Brand: Thermo Scientific Alfa Aesar H61089.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 187604-75-7
Molecular Formula C8H8BrNS
Molecular Weight (g/mol) 230.123
MDL Number MFCD23379863
InChI Key AUIBYERXYVUBIL-UHFFFAOYSA-N
Synonym 6-bromo-3,4-dihydro-2h-benzo b 1,4 thiazine
PubChem CID 67172430
IUPAC Name 6-bromo-3,4-dihydro-2H-1,4-benzothiazine
SMILES C1CSC2=C(N1)C=C(C=C2)Br

Specifications

Quantity 250 mg
Formula Weight 230.12
Percent Purity 97%
Chemical Name or Material 6-Bromo-3,4-dihydro-2H-1,4-benzothiazine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberI13Z035 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog NumberH61089.03,H61089.MD
Lot NumberJ10Z032 Certificate TypeCertificate of Analysis Date14/01/2025 Catalog NumberH61089.03,H61089.MD
Lot NumberX18X030 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog NumberH61089.03,H61089.MD
Lot NumberX24J020 Certificate TypeCertificate of Analysis Date09/11/2023 Catalog NumberH61089.03,H61089.MD
Lot NumberJ10Z032 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH61089.03,H61089.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6-Bromo-3,4-dihydro-2H-1,4-benzothiazine, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.