missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid hydrate, 95%, Thermo Scientific™
£67.80
Chemical Identifiers
CAS | 749849-14-7 |
---|---|
Molecular Formula | C8H5BrN2O2 |
Molecular Weight (g/mol) | 241.04 |
MDL Number | MFCD04116918 |
InChI Key | FQBFPVLZLNEQOE-UHFFFAOYSA-N |
Synonym | 6-bromoimidazo 1,2-a pyridine-2-carboxylic acid, 6-bromo-imidazo 1,2-a pyridine-2-carboxylic acid, imidazo 1,2-a pyridine-2-carboxylic acid, 6-bromo, 6-bromo-4-hydroimidazo 1,2-a pyridine-2-carboxylic acid, pubchem15865, buttpark 154\50-31, emcs ;6-maleimidocaproic acid nhs, 6-bromo-2-imidazo 1,2-a pyridinecarboxylic acid, 6-bromoimidazo 1,2-a pyridine-2-carboxylicacid |
PubChem CID | 2757018 |
IUPAC Name | 6-bromoimidazo[1,2-a]pyridine-2-carboxylic acid |
SMILES | OC(=O)C1=CN2C=C(Br)C=CC2=N1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15419045
|
Thermo Scientific Alfa Aesar
H35790.03 |
1 g | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
749849-14-7 | |
241.04 | |
FQBFPVLZLNEQOE-UHFFFAOYSA-N | |
2757018 | |
OC(=O)C1=CN2C=C(Br)C=CC2=N1 |
C8H5BrN2O2 | |
MFCD04116918 | |
6-bromoimidazo 1,2-a pyridine-2-carboxylic acid, 6-bromo-imidazo 1,2-a pyridine-2-carboxylic acid, imidazo 1,2-a pyridine-2-carboxylic acid, 6-bromo, 6-bromo-4-hydroimidazo 1,2-a pyridine-2-carboxylic acid, pubchem15865, buttpark 154\50-31, emcs ;6-maleimidocaproic acid nhs, 6-bromo-2-imidazo 1,2-a pyridinecarboxylic acid, 6-bromoimidazo 1,2-a pyridine-2-carboxylicacid | |
6-bromoimidazo[1,2-a]pyridine-2-carboxylic acid |
Specifications
749849-14-7 | |
MFCD04116918 | |
FQBFPVLZLNEQOE-UHFFFAOYSA-N | |
6-bromoimidazo[1,2-a]pyridine-2-carboxylic acid | |
2757018 | |
95% |
C8H5BrN2O2 | |
6-bromoimidazo 1,2-a pyridine-2-carboxylic acid, 6-bromo-imidazo 1,2-a pyridine-2-carboxylic acid, imidazo 1,2-a pyridine-2-carboxylic acid, 6-bromo, 6-bromo-4-hydroimidazo 1,2-a pyridine-2-carboxylic acid, pubchem15865, buttpark 154\50-31, emcs ;6-maleimidocaproic acid nhs, 6-bromo-2-imidazo 1,2-a pyridinecarboxylic acid, 6-bromoimidazo 1,2-a pyridine-2-carboxylicacid | |
OC(=O)C1=CN2C=C(Br)C=CC2=N1 | |
241.04 | |
241.05 (Anhydrous) | |
6-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid hydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures