missing translation for 'onlineSavingsMsg'
Learn More
6-Bromoimidazo[1,2-a]pyridine-3-carboxylic acid, 97%, Thermo Scientific™ Product Code.: 15532303

6-Bromoimidazo[1,2-a]pyridine-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 15532303
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15532303

Brand: Thermo Scientific Acros 449910050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 944896-42-8
Molecular Formula C8H5BrN2O2
Molecular Weight (g/mol) 241.04
MDL Number MFCD10698063
InChI Key SDKJLYHPCBMDAD-UHFFFAOYSA-N
Synonym 6-bromoimidazo 1,2-a pyridine-3-carboxylic acid, 6-bromoimidazo 1,2-a pyridine-3-carboxylicacid, 6-bromoh-imidazo 1,2-a pyridine-3-carboxylic acid, 6-bromo-1h-imidazo 1,2-a pyridine-3-carboxylic acid, 6-bromo-3-carboxyimidazo 1,2-a pyridine, 6-bromo-imidazo 1,2-a pyridine-3-carboxylic acid, imidazo 1,2-a pyridine-3-carboxylic acid, 6-bromo
PubChem CID 22031912
IUPAC Name 6-bromoimidazo[1,2-a]pyridine-3-carboxylic acid
SMILES OC(=O)C1=CN=C2C=CC(Br)=CN12

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 241.04
Percent Purity 97%
Chemical Name or Material 6-Bromoimidazo[1,2-a]pyridine-3-carboxylic acid
compliance-icons
Product Identifier
  • 6-Bromoimidazo¢1,2-a!pyridine-3-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6-Bromoimidazo[1,2-a]pyridine-3-carboxylic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.