Learn More
6-Bromoimidazo[1,2-a]pyridine, 98%, Thermo Scientific Chemicals
Description
It is used in organic syntheses and as pharmaceutical intermediates. The best results were obtained with lactams that could be introduced on the 6-bromoimidazo[1,2-a]pyridine. 1,2-dihydro-5-imidazo [1,2-a]pyridin-6-yl-6-methyl-2-oxo-3-pyridinecarbonitrile hydrochloride monohydrate 6 was synthesized in 5 steps from 6-bromoimidazo[1,2-a] pyridine using [2- 14 C] cyanoacetamide as the source of the radiolabel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is used in organic syntheses and as pharmaceutical intermediates. The best results were obtained with lactams that could be introduced on the 6-bromoimidazo[1,2-a]pyridine. 1,2-dihydro-5-imidazo [1,2-a]pyridin-6-yl-6-methyl-2-oxo-3-pyridinecarbonitrile hydrochloride monohydrate 6 was synthesized in 5 steps from 6-bromoimidazo[1,2-a] pyridine using [2- 14 C] cyanoacetamide as the source of the radiolabel.
Solubility
slightly soluble in water.
Notes
Incompatible with strong oxidizing agents. Store at room temperature.

Specifications
Specifications
Melting Point | 78°C to 80°C |
Quantity | 1 g |
Solubility Information | slightly soluble in water. |
Formula Weight | 197.03 |
Percent Purity | 98% |
Chemical Name or Material | 6-Bromoimidazo[1,2-a]pyridine |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.