missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-(Bromomethyl)-4-chloro-2-(trifluoromethyl)quinoline, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 6-bromomethyl-4-chloro-2-trifluoromethyl quinoline, quinoline,6-bromomethyl-4-chloro-2-trifluoromethyl, quinoline, 6-bromomethyl-4-chloro-2-trifluoromethyl, pubchem6004, acmc-1bzc1, buttpark 29\04-10, 6-bromomethyl-4-chloro-2-trifluoromethyl-quinoline, 2-trifluoromethyl-4-chloro-6-bromomethyl quinoline |
Formula Weight | 324.53 |
Percent Purity | 95% |
Chemical Name or Material | 6-(Bromomethyl)-4-chloro-2-(trifluoromethyl)quinoline |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
6-(Bromomethyl)-4-chloro-2-(trifluoromethyl)quinoline, ≥95%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction