missing translation for 'onlineSavingsMsg'
Learn More

6-chloro-1-hexyne, 97%, Thermo Scientific Chemicals

£43.32 - £148.74

Chemical Identifiers

CAS 10297-06-0
Molecular Formula C6H9Cl
Molecular Weight (g/mol) 116.59
MDL Number MFCD00013697
InChI Key ZUKOCGMVJUXIJA-UHFFFAOYSA-N
PubChem CID 272001
IUPAC Name 6-chlorohex-1-yne
SMILES ClCCCCC#C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10341223
View Documents Promotion Details
Thermo Scientific Acros
380810050
5 mL Glass bottle
£43.32
5mL
Please to purchase this item. Need a web account? Register with us today!
10301273
View Documents
Thermo Scientific Acros
380810250
25 mL Glass bottle
£148.74
25mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10297-06-0
116.59
ZUKOCGMVJUXIJA-UHFFFAOYSA-N
6-chlorohex-1-yne
C6H9Cl
MFCD00013697
272001
ClCCCCC#C

Specifications

10297-06-0
100.0
0.9620g/mL
37°C
96% min. (GC)
CH∽C(CH2)4Cl
MFCD00013697
6-chloro-1-hexyne, 5-hexynechloride, 1-hexyne,6-chloro, 1-hexyne, 6-chloro, 5-hexynyl chloride, 1-chloro-5-hexyne, 6-chloro-hex-1-yne, acmc-1boe0, zukocgmvjuxija-uhfffaoysa
ClCCCCC#C
116.59
116.59
Liquid
96.0
Yellow
131°C
Authentic
C6H9Cl
1.4490 to 1.451
0.962
ZUKOCGMVJUXIJA-UHFFFAOYSA-N
6-chlorohex-1-yne
272001
97%
6-Chloro-1-hexyne
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Product Identifier
  • 6-Chloro-1-hexyne

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.