missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2,3-difluorophenol, 97%, Thermo Scientific™

Product Code. 15456757
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 186590-18-1
Molecular Formula C6H3ClF2O
Molecular Weight (g/mol) 164.536
MDL Number MFCD22201050
InChI Key LFTQGYACSLFIRK-UHFFFAOYSA-N
Synonym phenol, 6-chloro-2,3-difluoro
PubChem CID 45119168
IUPAC Name 6-chloro-2,3-difluorophenol
SMILES C1=CC(=C(C(=C1F)F)O)Cl

Specifications

Quantity 1 g
Formula Weight 164.54
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 6-Chloro-2,3-difluorophenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'