missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2-methylpyridine-3-boronic acid, 98%, Thermo Scientific™

Product Code. 15449927
Click to view available options
Quantity:
250 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 913836-15-4
Molecular Formula C6H7BClNO2
Molecular Weight (g/mol) 171.387
MDL Number MFCD07368875
InChI Key WEKRUIAZOLIAOG-UHFFFAOYSA-N
Synonym 6-chloro-2-methylpyridin-3-yl boronic acid, 6-chloro-2-methylpyridine-3-boronic acid, 6-chloro-2-methylpyridine-3-boronicacid, 6-chloro-2-picoline-3-boronic acid, 6-chloro-2-methyl-3-pyridineboronic acid, boronicacid, b-6-chloro-2-methyl-3-pyridinyl, b-6-chloro-2-methyl-3-pyridinyl-boronic acid, pubchem1735, acmc-209raz, ksc486i0r
PubChem CID 44119152
IUPAC Name (6-chloro-2-methylpyridin-3-yl)boronic acid
SMILES B(C1=C(N=C(C=C1)Cl)C)(O)O

Specifications

Melting Point 154°C to 156°C
Quantity 250 mg
Formula Weight 171.39
Percent Purity 98%
Chemical Name or Material 6-Chloro-2-methylpyridine-3-boronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.