missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2H-chromene-3-carbonyl chloride, 95%, Thermo Scientific™

Product Code. 10140432
1 g
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 306935-54-6
Molecular Formula C10H6Cl2O2
Molecular Weight (g/mol) 229.056
MDL Number MFCD02091502
InChI Key HGFGWTNRUGRBNQ-UHFFFAOYSA-N
Synonym 6-chloro-2h-1-benzopyran-3-carbonyl chloride, 2h-1-benzopyran-3-carbonylchloride, 6-chloro, 6-chloro-2h-1-benzopyran-3-carbonylchloride, 6-chloranyl-2h-chromene-3-carbonyl chloride
PubChem CID 2735716
IUPAC Name 6-chloro-2H-chromene-3-carbonyl chloride
SMILES C1C(=CC2=C(O1)C=CC(=C2)Cl)C(=O)Cl

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Quantity 1 g
Formula Weight 229.06
Percent Purity 95%
Chemical Name or Material 6-Chloro-2H-chromene-3-carbonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6-Chloro-2H-chromene-3-carbonyl chloride, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.