missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-3(2H)-pyridazinone, 98%, Thermo Scientific Chemicals

Product Code. 15468418
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19064-67-6
Molecular Formula C4H3ClN2O
Molecular Weight (g/mol) 130.531
MDL Number MFCD00160460
InChI Key YICPBKWYZXFJNB-UHFFFAOYSA-N
Synonym 6-chloropyridazin-3-ol, 6-chloro-3-hydroxypyridazine, 6-chloro-2h-pyridazin-3-one, 6-chloropyridazin-3 2h-one, 3-chloro-6-hydroxypyridazine, 3 2h-pyridazinone, 6-chloro, 6-chloro-3 2h-pyridazinone, 3-chloro-6-pyridazone, 3-hydroxy-6-chloropyridazine, 6-chloro-3-pyridazinol
PubChem CID 252828
IUPAC Name 3-chloro-1H-pyridazin-6-one
SMILES C1=CC(=NNC1=O)Cl

Specifications

Quantity 1 g
Formula Weight 130.53
Percent Purity 98%
Chemical Name or Material 6-Chloro-3(2H)-pyridazinone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'