missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-4-methylpyridine-2-carboxylic acid, 95%, Thermo Scientific Chemicals

Product Code. 15460349
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 324028-95-7
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD09834827
InChI Key IDIRHQNTRMGBPJ-UHFFFAOYSA-N
Synonym 6-chloro-4-methyl-2-pyridinecarboxylic acid, 6-chloro-4-methylpicolinic acid, 6-chloro-4-methyl-pyridine-2-carboxylic acid, 6-chloranyl-4-methyl-pyridine-2-carboxylic acid
PubChem CID 23002381
IUPAC Name 6-chloro-4-methylpyridine-2-carboxylic acid
SMILES CC1=CC(=NC(=C1)Cl)C(=O)O

Specifications

Color White
Quantity 5 g
UN Number UN2811
Formula Weight 171.58
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 6-Chloro-4-methylpyridine-2-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'