Learn More
Thermo Scientific Maybridge 6-Chlorobenzo[d]isoxazol-3-ol, 1g, 97%
Brand: Thermo Scientific Maybridge RF01288DA
6-Chlorobenzo[d]isoxazol-3-ol, 1g, 97%
Code : 31
Additional Details : CAS Number : 61977-29-5 Weight : 0.05100kg
Brand: Thermo Scientific Maybridge RF01288DA
6-Chlorobenzo[d]isoxazol-3-ol, 1g, 97%
Code : 31
Additional Details : CAS Number : 61977-29-5 Weight : 0.05100kg