missing translation for 'onlineSavingsMsg'
Learn More

6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate, 95%

Product Code. 15489025
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15489025

Brand: Thermo Scientific Alfa Aesar H35567.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 138642-97-4 | C8H5ClN2O2 | 196.59 g/mol

6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138642-97-4
Molecular Formula C8H5ClN2O2
Molecular Weight (g/mol) 196.59
MDL Number MFCD07021509
InChI Key LXRDUEDQFSBROM-UHFFFAOYSA-N
Synonym 6-chloroimidazo 1,2-a pyridine-3-carboxylic acid, imidazo 1,2-a pyridine-3-carboxylic acid, 6-chloro
PubChem CID 16413639
IUPAC Name 6-chloroimidazo[1,2-a]pyridine-3-carboxylic acid
SMILES OC(=O)C1=CN=C2C=CC(Cl)=CN12

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 196.59 (Anhydrous)
Percent Purity 95%
Chemical Name or Material 6-Chloroimidazo[1,2-a]pyridine-3-carboxylic acid hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.