missing translation for 'onlineSavingsMsg'
Learn More

6-Chloroindole, 99%, Thermo Scientific™

£29.20 - £70.30

Chemical Identifiers

CAS 17422-33-2
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.59
MDL Number MFCD00005681
InChI Key YTYIMDRWPTUAHP-UHFFFAOYSA-N
Synonym 6-chloroindole, 1h-indole, 6-chloro, 6-chloro indole, 6-chloro-indole, 6-chlor-1h-indole, pubchem1669, 6-chloro-1-h-indole, #, ksc174m9p
PubChem CID 87111
ChEBI CHEBI:80918
IUPAC Name 6-chloro-1H-indole
SMILES ClC1=CC=C2C=CNC2=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10245541
View Documents
Thermo Scientific Acros
331622500
250 mg Glass bottle
£29.20
250mg
Please to purchase this item. Need a web account? Register with us today!
10777792
View Documents
Thermo Scientific Acros
331620010
1 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

17422-33-2
151.59
YTYIMDRWPTUAHP-UHFFFAOYSA-N
87111
6-chloro-1H-indole
C8H6ClN
MFCD00005681
6-chloroindole, 1h-indole, 6-chloro, 6-chloro indole, 6-chloro-indole, 6-chlor-1h-indole, pubchem1669, 6-chloro-1-h-indole, #, ksc174m9p
CHEBI:80918
ClC1=CC=C2C=CNC2=C1

Specifications

17422-33-2
100.0
Beige
98.5% min. (GC)
MFCD00005681
Solubility in water: slightly soluble
ClC1=CC=C2C=CNC2=C1
151.59
CHEBI:80918
99%
6-Chloroindole, 99%
98.5
88°C to 91°C
Authentic
C8H6ClN
6-chloroindole, 1h-indole, 6-chloro, 6-chloro indole, 6-chloro-indole, 6-chlor-1h-indole, pubchem1669, 6-chloro-1-h-indole, #, ksc174m9p
YTYIMDRWPTUAHP-UHFFFAOYSA-N
6-chloro-1H-indole
87111
151.6
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 241-449-4

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only