Learn More
6-chloropyridazine-3-carboxylic acid, 97%, Thermo Scientific™
£127.00
Chemical Identifiers
CAS | 5096-73-1 |
---|---|
Molecular Formula | C5H3ClN2O2 |
Molecular Weight (g/mol) | 158.54 |
MDL Number | MFCD00160464 |
InChI Key | HHGZQZULOHYEOH-UHFFFAOYSA-N |
Synonym | 6-chloro-pyridazine-3-carboxylic acid, 3-chloropyridazine-6-carboxylic acid, 3-pyridazinecarboxylic acid, 6-chloro, 6-chloro-3-carboxy-pyridazine, 3-carboxy-6-chloropyridazine, 6-chloro-3-pyridazine-carboxylic acid, 6-chloropyridazine-3-carboxylic, 6-chloropyridazine-3-carboxylicacid, pubchem17692 |
PubChem CID | 6415762 |
IUPAC Name | 6-chloropyridazine-3-carboxylic acid |
SMILES | OC(=O)C1=NN=C(Cl)C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10411538
|
Thermo Scientific Acros
429530010 |
1 g | Glass bottle | N/A | |||||
Description
Chemical Identifiers
5096-73-1 | |
158.54 | |
HHGZQZULOHYEOH-UHFFFAOYSA-N | |
6415762 | |
OC(=O)C1=NN=C(Cl)C=C1 |
C5H3ClN2O2 | |
MFCD00160464 | |
6-chloro-pyridazine-3-carboxylic acid, 3-chloropyridazine-6-carboxylic acid, 3-pyridazinecarboxylic acid, 6-chloro, 6-chloro-3-carboxy-pyridazine, 3-carboxy-6-chloropyridazine, 6-chloro-3-pyridazine-carboxylic acid, 6-chloropyridazine-3-carboxylic, 6-chloropyridazine-3-carboxylicacid, pubchem17692 | |
6-chloropyridazine-3-carboxylic acid |
Specifications
5096-73-1 | |
100.0 | |
White to Cream | |
96% min. (HPLC) | |
MFCD00160464 | |
HHGZQZULOHYEOH-UHFFFAOYSA-N | |
6-chloropyridazine-3-carboxylic acid | |
6415762 | |
97% | |
6-chloropyridazine-3-carboxylic acid |
96.0 | |
145.0°C to 148.0°C | |
Authentic | |
C5H3ClN2O2 | |
6-chloro-pyridazine-3-carboxylic acid, 3-chloropyridazine-6-carboxylic acid, 3-pyridazinecarboxylic acid, 6-chloro, 6-chloro-3-carboxy-pyridazine, 3-carboxy-6-chloropyridazine, 6-chloro-3-pyridazine-carboxylic acid, 6-chloropyridazine-3-carboxylic, 6-chloropyridazine-3-carboxylicacid, pubchem17692 | |
OC(=O)C1=NN=C(Cl)C=C1 | |
158.54 | |
158.54 | |
Crystalline Powder |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only