missing translation for 'onlineSavingsMsg'
Learn More

6-Chloropyridine-3-carbohydrazide, ≥95%, Thermo Scientific™

Product Code. 10365252
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10365252

Brand: Thermo Scientific Maybridge SPB07126DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 168893-66-1
Molecular Formula C6H6ClN3O
Molecular Weight (g/mol) 171.58
MDL Number MFCD00221429
InChI Key BIECNFYNFQQRRP-UHFFFAOYSA-N
Synonym 6-chloronicotinohydrazide, 2-chloropyridine-5-carbohydrazide, 6-chloronicotinic acid hydrazide, pubchem18551, buttpark 9157-08, 2-chloro-5-hydrazinocarbonyl pyridine, 6-chloropyridine-3-carbohydrazide, 95+%, 2-chloropyridine-5-carboxylic acid hydrazide, 3-pyridinecarboxylicacid, 6-chloro-, hydrazide, 3-pyridinecarboxylic acid, 6-chloro-, hydrazide
PubChem CID 2800177
IUPAC Name 6-chloropyridine-3-carbohydrazide
SMILES NNC(=O)C1=CN=C(Cl)C=C1

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 171.59
Percent Purity 95%
Chemical Name or Material 6-Chloropyridine-3-carbohydrazide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.