missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chloroquinoline, 98%, Thermo Scientific™
£25.14 - £100.24
Chemical Identifiers
CAS | 612-57-7 |
---|---|
Molecular Formula | C9H6ClN |
Molecular Weight (g/mol) | 163.60 |
MDL Number | MFCD00024024 |
InChI Key | GKJSZXGYFJBYRQ-UHFFFAOYSA-N |
Synonym | quinoline, 6-chloro, unii-k4a4bau5se, ccris 3981, k4a4bau5se, 6-chloro-quinoline, 6-chlor-chinolin;, zlchem 764, pubchem5813, acmc-1aulp, quinoline, 6-chloro-, |
PubChem CID | 69163 |
IUPAC Name | 6-chloroquinoline |
SMILES | ClC1=CC=C2N=CC=CC2=C1 |
Description
Chemical Identifiers
612-57-7 | |
163.60 | |
GKJSZXGYFJBYRQ-UHFFFAOYSA-N | |
69163 | |
ClC1=CC=C2N=CC=CC2=C1 |
C9H6ClN | |
MFCD00024024 | |
quinoline, 6-chloro, unii-k4a4bau5se, ccris 3981, k4a4bau5se, 6-chloro-quinoline, 6-chlor-chinolin;, zlchem 764, pubchem5813, acmc-1aulp, quinoline, 6-chloro-, | |
6-chloroquinoline |
Specifications
612-57-7 | |
1.2700g/mL | |
145°C | |
97.5% min. (GC) | |
C9H6ClN | |
1.27 | |
GKJSZXGYFJBYRQ-UHFFFAOYSA-N | |
6-chloroquinoline | |
69163 | |
98% |
39.0°C to 42.0°C | |
263.0°C | |
Authentic | |
Glass bottle | |
MFCD00024024 | |
quinoline, 6-chloro, unii-k4a4bau5se, ccris 3981, k4a4bau5se, 6-chloro-quinoline, 6-chlor-chinolin;, zlchem 764, pubchem5813, acmc-1aulp, quinoline, 6-chloro-, | |
ClC1=CC=C2N=CC=CC2=C1 | |
163.60 | |
163.61 | |
6-Chloroquinoline |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 210-314-1
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 6-Chloroquinoline, 98%
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only