missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoro-1,2,3,4-tetrahydroquinoline, 97%, Thermo Scientific Chemicals

£89.60 - £245.00

Chemical Identifiers

CAS 59611-52-8
Molecular Formula C9H10FN
Molecular Weight (g/mol) 151.18
MDL Number MFCD09040997
InChI Key NECDDBBJVCNJNS-UHFFFAOYSA-N
Synonym quinoline, 6-fluoro-1,2,3,4-tetrahydro, 6-fluoro-1,2,3,4-tetrahydro-quinoline
PubChem CID 14783911
IUPAC Name 6-fluoro-1,2,3,4-tetrahydroquinoline
SMILES FC1=CC2=C(NCCC2)C=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15437507
View Documents
Thermo Scientific Alfa Aesar
H35364.MD
250 mg
£89.60
250mg
Please to purchase this item. Need a web account? Register with us today!
15427507
View Documents
Thermo Scientific Alfa Aesar
H35364.03
1 g
£245.00
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

6-Fluoro-1,2,3,4-tetrahydroquinoline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Fluoro-1,2,3,4-tetrahydroquinoline is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

59611-52-8
151.18
NECDDBBJVCNJNS-UHFFFAOYSA-N
14783911
FC1=CC2=C(NCCC2)C=C1
C9H10FN
MFCD09040997
quinoline, 6-fluoro-1,2,3,4-tetrahydro, 6-fluoro-1,2,3,4-tetrahydro-quinoline
6-fluoro-1,2,3,4-tetrahydroquinoline

Specifications

59611-52-8
C9H10FN
quinoline, 6-fluoro-1,2,3,4-tetrahydro, 6-fluoro-1,2,3,4-tetrahydro-quinoline
FC1=CC2=C(NCCC2)C=C1
151.18
151.18
6-Fluoro-1,2,3,4-tetrahydroquinoline
32°C to 34°C
MFCD09040997
NECDDBBJVCNJNS-UHFFFAOYSA-N
6-fluoro-1,2,3,4-tetrahydroquinoline
14783911
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only