missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Fluoro-2,3-dihydro-4H-thiochromen-4-one, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Quantity | 5 g |
Formula Weight | 182.22 |
Percent Purity | 95% |
Chemical Name or Material | 6-Fluoro-2, 3-dihydro-4H-thiochromen-4-one |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
6-Fluoro-2,3-dihydro-4H-thiochromen-4-one, ≥95%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction