missing translation for 'onlineSavingsMsg'
Learn More

6-(Fmoc-amino)hexanoic acid, 98%, Thermo Scientific Chemicals

Product Code. 15479678
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

6-(Fmoc-amino)hexanoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-(Fmoc-amino)hexanoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in 1% acetic acid, water and 1mmol in 2mL DMF clearly soluble.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88574-06-5
Molecular Formula C21H23NO4
Molecular Weight (g/mol) 353.418
MDL Number MFCD00077045
InChI Key FPCPONSZWYDXRD-UHFFFAOYSA-N
Synonym fmoc-6-ahx-oh, fmoc-epsilon-acp-oh, n-fmoc-6-aminohexanoic acid, fmoc-, a-acp-oh, 6-9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid, 6-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, 6-9h-fluoren-9-ylmethoxycarbonylamino hexanoic acid, hexanoic acid, 6-9h-fluoren-9-ylmethoxy carbonyl amino
PubChem CID 2756087
IUPAC Name 6-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCCCC(=O)O

Specifications

Color White
Linear Formula FmocNH(CH2)5CO2H
Quantity 5 g
Beilstein 5883220
Solubility Information Soluble in 1% acetic acid,water and 1mmol in 2mL DMF clearly soluble.
Formula Weight 353.41
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 6-(Fmoc-amino)hexanoic acid
compliance-icons
Product Identifier
  • 6-(Fmoc-amino)hexanoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 6-(FMOC-Amino)hexanoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.