missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-(Fmoc-amino)hexanoic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
6-(Fmoc-amino)hexanoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-(Fmoc-amino)hexanoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in 1% acetic acid, water and 1mmol in 2mL DMF clearly soluble.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
6-(Fmoc-amino)hexanoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in 1% acetic acid, water and 1mmol in 2mL DMF clearly soluble.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | White |
Linear Formula | FmocNH(CH2)5CO2H |
Quantity | 25 g |
Beilstein | 5883220 |
Solubility Information | Soluble in 1% acetic acid,water and 1mmol in 2mL DMF clearly soluble. |
Formula Weight | 353.41 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | 6-(Fmoc-amino)hexanoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'