missing translation for 'onlineSavingsMsg'
Learn More

6-Hydrazino-1,3,4-trimethyl-1H-pyrazolo[3,4-b]pyridine, 97%, Thermo Scientific™

Product Code. 10672633
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10672633

Brand: Thermo Scientific Maybridge KM02421CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 175202-00-3
Molecular Formula C9H13N5
Molecular Weight (g/mol) 191.24
MDL Number MFCD00067969
InChI Key NDFXPLOECYNSHX-UHFFFAOYSA-N
Synonym 6-hydrazino-1,3,4-trimethyl-1h-pyrazolo 3,4-b pyridine, 6-hydrazinyl-1,3,4-trimethyl-1h-pyrazolo 3,4-b pyridine, 6-hydrazinyl-1,3,4-trimethylpyrazolo 3,4-b pyridine, 1,3,4-trimethylpyrazolo 5,4-b pyridine-6-ylhydrazine, 1,3,4-trimethylpyrazolo 3,4-b pyridin-6-yl hydrazine, 1h-pyrazolo 3,4-b pyridine,6-hydrazinyl-1,3,4-trimethyl, 6h-pyrazolo 3,4-b pyridin-6-one, 1,7-dihydro-1,3,4-trimethyl-, hydrazone 9ci
PubChem CID 2778646
IUPAC Name 6-hydrazinyl-1,3,4-trimethyl-1H-pyrazolo[3,4-b]pyridine
SMILES CN1N=C(C)C2=C(C)C=C(NN)N=C12

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 191.24
Percent Purity 97%
Chemical Name or Material 6-Hydrazino-1,3,4-trimethyl-1H-pyrazolo[3,4-b]pyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.