Learn More
6-Hydroxyindole, 97%, Thermo Scientific Chemicals
£113.00 - £1330.76
Chemical Identifiers
CAS | 2380-86-1 |
---|---|
Molecular Formula | C8H7NO |
Molecular Weight (g/mol) | 133.15 |
InChI Key | XAWPKHNOFIWWNZ-UHFFFAOYSA-N |
Synonym | 6-hydroxyindole, 6-indolol, indolol, 6-hydroxy indole, unii-3i03jz599t, 6-hydroxy-1h-indole, 6hydroxyindole, 6-hydroxy-indole, indol-6-ol, 1h-indole-6-ol |
PubChem CID | 524508 |
IUPAC Name | 1H-indol-6-ol |
SMILES | C1=CC(=CC2=C1C=CN2)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10143712
|
Thermo Scientific Acros
379010010 |
1 g | Glass bottle |
£113.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10200163
|
Thermo Scientific Acros
379010050 |
5 g | Glass bottle |
£389.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11966631
|
Thermo Scientific Acros
379010250 |
25 g | Glass Bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2380-86-1 | |
133.15 | |
6-hydroxyindole, 6-indolol, indolol, 6-hydroxy indole, unii-3i03jz599t, 6-hydroxy-1h-indole, 6hydroxyindole, 6-hydroxy-indole, indol-6-ol, 1h-indole-6-ol | |
1H-indol-6-ol |
C8H7NO | |
XAWPKHNOFIWWNZ-UHFFFAOYSA-N | |
524508 | |
C1=CC(=CC2=C1C=CN2)O |
Specifications
2380-86-1 | |
White | |
97.5% min. (HPLC) | |
6-hydroxyindole, 6-indolol, indolol, 6-hydroxy indole, unii-3i03jz599t, 6-hydroxy-1h-indole, 6hydroxyindole, 6-hydroxy-indole, indol-6-ol, 1h-indole-6-ol | |
C1=CC(=CC2=C1C=CN2)O | |
133.15 | |
133.15 | |
Shiny Crystalline Powder |
124.0°C to 130.0°C | |
Authentic | |
C8H7NO | |
XAWPKHNOFIWWNZ-UHFFFAOYSA-N | |
1H-indol-6-ol | |
524508 | |
97% | |
6-Hydroxyindole |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protecti
GHS Signal Word: Danger
Recommended Storage : Product may darken during storage
- 6-Hydroxyindole
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H411-Toxic to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only