missing translation for 'onlineSavingsMsg'
Learn More

6-Iodo-2-methyl-1,3-benzothiazole, 90%, Thermo Scientific™

£35.70 - £266.00

Chemical Identifiers

CAS 68867-20-9
Molecular Formula C8H6INS
Molecular Weight (g/mol) 275.107
InChI Key YVXGKAZJAUWMPM-UHFFFAOYSA-N
Synonym 6-iodo-2-methyl-benzothiazole, 6-iodo-2-methylbenzo d thiazole, 2-methyl-6-iodobenzothiazole, 6-iodo-2-methylbenzothiazole, benzothiazole, 6-iodo-2-methyl, benzothiazole,6-iodo-2-methyl
PubChem CID 807592
IUPAC Name 6-iodo-2-methyl-1,3-benzothiazole
SMILES CC1=NC2=C(S1)C=C(C=C2)I
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10193364
View Documents
Thermo Scientific Maybridge
CC54711CB
250 mg
£35.70
250mg
Please to purchase this item. Need a web account? Register with us today!
10546324
View Documents
Thermo Scientific Maybridge
CC54711DA
1 g
£88.60
1g
Please to purchase this item. Need a web account? Register with us today!
10103374
View Documents
Thermo Scientific Maybridge
CC54711DE
5 g
£266.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

68867-20-9
275.107
6-iodo-2-methyl-benzothiazole, 6-iodo-2-methylbenzo d thiazole, 2-methyl-6-iodobenzothiazole, 6-iodo-2-methylbenzothiazole, benzothiazole, 6-iodo-2-methyl, benzothiazole,6-iodo-2-methyl
6-iodo-2-methyl-1,3-benzothiazole
C8H6INS
YVXGKAZJAUWMPM-UHFFFAOYSA-N
807592
CC1=NC2=C(S1)C=C(C=C2)I

Specifications

68867-20-9
C8H6INS
YVXGKAZJAUWMPM-UHFFFAOYSA-N
6-iodo-2-methyl-1,3-benzothiazole
807592
90%
90%
6-iodo-2-methyl-benzothiazole, 6-iodo-2-methylbenzo d thiazole, 2-methyl-6-iodobenzothiazole, 6-iodo-2-methylbenzothiazole, benzothiazole, 6-iodo-2-methyl, benzothiazole,6-iodo-2-methyl
CC1=NC2=C(S1)C=C(C=C2)I
275.107
275.11
6-Iodo-2-methyl-1, 3-benzothiazole
Videos
SDS
Documents

Documents

Product Certifications