missing translation for 'onlineSavingsMsg'
Learn More

6-Mercaptopurine monohydrate, 98%, Thermo Scientific Chemicals

Product Code. 10761501
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6112-76-1
Molecular Formula C5H6N4OS
Molecular Weight (g/mol) 170.19
MDL Number MFCD03854445,MFCD01461928
InChI Key WFFQYWAAEWLHJC-UHFFFAOYSA-N
Synonym 6-mercaptopurine monohydrate, 6-mercaptopurine hydrate, mercaptopurine monohydrate, mercaptopurine hydrate, 6h-purine-6-thione, 1,7-dihydro-, monohydrate, 6,7-dihydro-3h-purine-6-thione hydrate, purine-6-thiol monohydrate, 1,7-dihydro-6h-purine-6-thione monohydrate, purine-6-thiol, monohydrate, unii-e7wed276i5
PubChem CID 2724350
ChEBI CHEBI:31822
IUPAC Name 3,7-dihydropurine-6-thione;hydrate
SMILES O.S=C1N=CNC2=C1NC=N2

Specifications

CAS Min % 99.5
CAS Max % 100.0
Melting Point 313.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Merck Index 10, 5702
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in diluted alkali solutions,slightly soluble in diluted sulfuric acid,soluble in hot alcohol,insoluble in acetone and ether
Formula Weight 170.19
Percent Purity 99.5+%
Physical Form Fine Powder
Chemical Name or Material 6-Mercaptopurine monohydrate, 99.5+%
Show More Show Less
compliance-icons
Product Identifier
  • 6-Mercaptopurine monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'