Learn More
6-Methoxy-1,2,3,4-tetrahydroquinoline, 97%, Thermo Scientific Chemicals
£25.63 - £229.00
Chemical Identifiers
CAS | 120-15-0 |
---|---|
Molecular Formula | C10H13NO |
Molecular Weight (g/mol) | 163.22 |
MDL Number | MFCD00090494 |
InChI Key | FRXSZNDVFUDTIR-UHFFFAOYSA-N |
Synonym | thallin, thalline, 6-methoxy-1,2,3,4-tetrahydro-quinoline, 6-methoxytetrahydroquinoline, quinoline, 1,2,3,4-tetrahydro-6-methoxy, 1,2,3,4-tetrahydro-6-methoxyquinoline, methyl 1,2,3,4-tetrahydro-6-quinolyl ether, quinoline l,2,3,4-tetrahydro-6-methoxy, wln: t66 bmt & j ho1, 1,3,4-tetrahydro-6-methoxyquinoline |
PubChem CID | 67113 |
IUPAC Name | 6-methoxy-1,2,3,4-tetrahydroquinoline |
SMILES | COC1=CC2=C(C=C1)NCCC2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15479428
|
Thermo Scientific Alfa Aesar
H63002.MD |
250 mg |
£25.63
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15459428
|
Thermo Scientific Alfa Aesar
H63002.03 |
1 g |
£61.20
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15469428
|
Thermo Scientific Alfa Aesar
H63002.06 |
5 g |
£229.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,2,3,4-Tetrahydro-6-methoxyquinoline is used as chemical reagents, organic intermediates, fine chemicals, pharmaceutical research and development.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,2,3,4-Tetrahydro-6-methoxyquinoline is used as chemical reagents, organic intermediates, fine chemicals, pharmaceutical research and development.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
120-15-0 | |
163.22 | |
FRXSZNDVFUDTIR-UHFFFAOYSA-N | |
67113 | |
COC1=CC2=C(C=C1)NCCC2 |
C10H13NO | |
MFCD00090494 | |
thallin, thalline, 6-methoxy-1,2,3,4-tetrahydro-quinoline, 6-methoxytetrahydroquinoline, quinoline, 1,2,3,4-tetrahydro-6-methoxy, 1,2,3,4-tetrahydro-6-methoxyquinoline, methyl 1,2,3,4-tetrahydro-6-quinolyl ether, quinoline l,2,3,4-tetrahydro-6-methoxy, wln: t66 bmt & j ho1, 1,3,4-tetrahydro-6-methoxyquinoline | |
6-methoxy-1,2,3,4-tetrahydroquinoline |
Specifications
120-15-0 | |
C10H13NO | |
thallin, thalline, 6-methoxy-1,2,3,4-tetrahydro-quinoline, 6-methoxytetrahydroquinoline, quinoline, 1,2,3,4-tetrahydro-6-methoxy, 1,2,3,4-tetrahydro-6-methoxyquinoline, methyl 1,2,3,4-tetrahydro-6-quinolyl ether, quinoline l,2,3,4-tetrahydro-6-methoxy, wln: t66 bmt & j ho1, 1,3,4-tetrahydro-6-methoxyquinoline | |
COC1=CC2=C(C=C1)NCCC2 | |
163.22 | |
163.22 | |
6-Methoxy-1,2,3,4-tetrahydroquinoline |
34°C to 37°C | |
MFCD00090494 | |
FRXSZNDVFUDTIR-UHFFFAOYSA-N | |
6-methoxy-1,2,3,4-tetrahydroquinoline | |
67113 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 204-374-8
missing translation for 'rtecsNumber' : VC2940000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only