missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-Methoxy-2-naphthylmagnesium bromide, 0.5M in THF

Product Code. 15400736
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 38046-82-1
Molecular Formula C11H9BrMgO
MDL Number MFCD01311488
Quantity 100 mL
UN Number UN2924
Synonym 6-methoxy-2-naphthylmagnesium bromide, sogqgmfueumyjd-uhfffaoysa-m, 6-methoxynaphthalen-2-ylmagnesium bromide, 4-fluorobenzylmagnesium chloride 0.25m&, 6-methoxy-2-naphthylmagnesium bromide, 0.5m in thf, 6-methoxy-2-naphthylmagnesium bromide 0.5 m in tetrahydrofuran, 6-methoxy-2-naphthylmagnesium bromide solution, 0.5 m in thf
InChI Key SOGQGMFUEUMYJD-UHFFFAOYSA-M
SMILES COC1=CC2=CC=C([Mg]Br)C=C2C=C1
IUPAC Name magnesium;6-methoxy-2H-naphthalen-2-ide;bromide
Molecular Weight (g/mol) 261.40
PubChem CID 11242453
Formula Weight 261.4
Concentration or Composition (by Analyte or Components) 0.5M in THF
Chemical Name or Material 6-Methoxy-2-naphthylmagnesium bromide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'