missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxynicotinonitrile, 97%, Thermo Scientific™

Product Code. 10477442
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
This item is not returnable. View return policy

Product Code. 10477442

Brand: Thermo Scientific Maybridge SPB04785DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 15871-85-9
Molecular Formula C7H6N2O
Molecular Weight (g/mol) 134.138
MDL Number MFCD00084981
InChI Key DFPYAQAFVHRSAG-UHFFFAOYSA-N
Synonym 6-methoxynicotinonitrile, 2-methoxypyridine-5-carbonitrile, 6-methoxy-3-pyridinecarbonitrile, 2-methoxy-5-cyanopyridine, 5-cyano-2-methoxypyridine, 2-methoxy-5-pyridinecarbonitrile, 6-methoxy-nicotinonitrile, 3-pyridinecarbonitrile, 6-methoxy, 6-metoxynicotinonitrile, pubchem15154
PubChem CID 2736776
IUPAC Name 6-methoxypyridine-3-carbonitrile
SMILES COC1=NC=C(C=C1)C#N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 134.14
Percent Purity 97%
Chemical Name or Material 6-Methoxynicotinonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.