missing translation for 'onlineSavingsMsg'
Learn More
6-Methyl-2-thiouracil, 98%, Thermo Scientific Chemicals Product Code.: 11469116

6-Methyl-2-thiouracil, 98%, Thermo Scientific Chemicals

Product Code. 11469116
250 g
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11469116

Brand: Thermo Scientific Alfa Aesar A17982.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

6-Methyl-2-thiouracil is mainly used in surgical preparation of mild hyperthyroidism, thyroid storm, hyperthyroidism and postoperative treatment. It is used in the treatment of hyperthyroidism. It is used as a drug, which is applied to decrease the formation of stored thyroid hormone and thyroglobulin in the thyroid gland.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methyl-2-thiouracil is mainly used in surgical preparation of mild hyperthyroidism, thyroid storm, hyperthyroidism and postoperative treatment. It is used in the treatment of hyperthyroidism. It is used as a drug, which is applied to decrease the formation of stored thyroid hormone and thyroglobulin in the thyroid gland.

Solubility
Soluble in ethanol. Slightly soluble in methanol, acetone, diethyl ether, water, ammonia and alkali hydroxides. Insoluble in benzene and chloroform.

Notes
Light sensitive. Incompatible with strong oxidizing agents. Keep the container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-04-2
Molecular Formula C5H6N2OS
Molecular Weight (g/mol) 142.18
MDL Number MFCD00006040
InChI Key HWGBHCRJGXAGEU-UHFFFAOYSA-N
Synonym methylthiouracil, 6-methyl-2-thiouracil, methiocil, 4-hydroxy-2-mercapto-6-methylpyrimidine, thimecil, thiomecil, thiothymin, alkiron, metacil, 2-mercapto-6-methylpyrimidin-4-olShow MoreShow Less
PubChem CID 667493
ChEBI CHEBI:82346
IUPAC Name 6-methyl-2-sulfanylidene-1H-pyrimidin-4-one
SMILES CC1=CC(=O)NC(=S)N1

Specifications

Melting Point ∼329°C (decomposition)
Quantity 250 g
Beilstein 115648
Merck Index 14,6128
Solubility Information Soluble in ethanol. Slightly soluble in methanol,acetone,diethyl ether,water,ammonia and alkali hydroxides. Insoluble in benzene and chloroform.
Formula Weight 142.18
Percent Purity 98%
Chemical Name or Material 6-Methyl-2-thiouracil

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN24A034 Certificate TypeCertificate of Analysis Date04/05/2024 Catalog NumberA17982.A1,A17982.18,A17982.30
Lot NumberS17B017 Certificate TypeCertificate of Analysis Date09/01/2024 Catalog NumberA17982.A1,A17982.18,A17982.30
Lot NumberX24I038 Certificate TypeCertificate of Analysis Date08/11/2022 Catalog NumberA17982.A1,A17982.18,A17982.30
Lot NumberY06B015 Certificate TypeCertificate of Analysis Date18/04/2022 Catalog NumberA17982.A1,A17982.18,A17982.30
Lot NumberF29U012 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17982.A1,A17982.18,A17982.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6-Methyl-2-thiouracil, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.