missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Methylchromone hydrate, 99%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Specifications
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | 76.0°C to 78.0°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass bottle |
Quantity | 5 g |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in chloroform,5% naoh solution,,ethylacetate,alcohol |
IUPAC Name | 6-methylchromen-4-one;hydrate |
Show More |
Safety and Handling
Product Identifier
- 6-Methylchromone hydrate
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'