missing translation for 'onlineSavingsMsg'
Learn More

6-Methylheptane-2,4-dione, 98+%

Product Code. p-7021151
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
This item is not returnable. View return policy

Product Code. 11374639

Brand: Thermo Scientific Alfa Aesar A13373.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3002-23-1 | C8H14O2 | 142.198 g/mol

6-Methylheptane-2,4-dione, is used as a multipurpose intermediate. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceutical industry

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methylheptane-2,4-dione, is used as a multipurpose intermediate. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceutical industry

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3002-23-1
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00008939
InChI Key IGMOYJSFRIASIE-UHFFFAOYSA-N
Synonym 6-methyl-2,4-heptanedione, isovalerylacetone, 2,4-heptanedione, 6-methyl, unii-o97vug08ub, 6-methylheptan-2,4-dione, o97vug08ub, nsc94, nsc 94, acmc-1ct2p, 6-methyl-2,4-heptandione
PubChem CID 76354
IUPAC Name 6-methylheptane-2,4-dione
SMILES CC(C)CC(=O)CC(=O)C

Specifications

Density 0.919
Boiling Point 183°C
Flash Point 58°C (136°F)
Refractive Index 1.456
Quantity 2 g
UN Number UN1224
Beilstein 1751122
Solubility Information It is insoluble in water.
Formula Weight 142.2
Percent Purity ≥98%
Chemical Name or Material 6-Methylheptane-2,4-dione
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.