missing translation for 'onlineSavingsMsg'
Learn More

6-Methylindole, 98%, Thermo Scientific Chemicals

Product Code. 11388466
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is a reactant for preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators, reactant for preparation of indole linked triazole derivatives as antifungal agents, reactant for preparation of N-β-D-xylosyl-indole derivatives as SGLT2 inhibitors for management of hyperglycemia in diabetes, reactant for preparation of aminoguanidine derivatives of arylsulfonylacylindoles as antifungal agents, reactant for preparation of indolylindazoles and indolylpyrazolopyridines as interleukin-2 inducible T cell kinase inhibitors, reactant for preparation of arylsulfonylacetylindoles as anti-HIV-1 agents. 6-Methylindole was used in the synthesis of benz[c,d]indol-3(1H)-one derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reactant for preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators, reactant for preparation of indole linked triazole derivatives as antifungal agents, reactant for preparation of N-β-D-xylosyl-indole derivatives as SGLT2 inhibitors for management of hyperglycemia in diabetes, reactant for preparation of aminoguanidine derivatives of arylsulfonylacylindoles as antifungal agents, reactant for preparation of indolylindazoles and indolylpyrazolopyridines as interleukin-2 inducible T cell kinase inhibitors, reactant for preparation of arylsulfonylacetylindoles as anti-HIV-1 agents. 6-Methylindole was used in the synthesis of benz[c,d]indol-3(1H)-one derivatives.

Solubility
Insoluble in water.

Notes
Moisture and light sensitive. Store in dry condition. Keep away from active metals, bases, oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3420-02-8
MDL Number MFCD00005682

Specifications

Melting Point 29°C to 32°C
Density 1.059
Boiling Point 112°C (5 mmHg)
Flash Point >110°C (230°F)
Molecular Formula C11H15NO2
Refractive Index 1.6071
Quantity 250 mg
Beilstein 109708
Sensitivity Light sensitive
Synonym n-boc-aniline, tert-butyl phenylcarbamate, n-boc aniline, t-butyl carbanilate, tert-butyl carbanilate, carbanilic acid, tert-butyl ester, carbamic acid, phenyl-, 1,1-dimethylethyl ester, n-tert-butoxycarbonyl aniline, tert-butyl n-phenylurethane, carbanilic acid tert-butyl ester
Solubility Information Insoluble in water.
InChI Key KZZHPWMVEVZEFG-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)NC1=CC=CC=C1
IUPAC Name tert-butyl N-phenylcarbamate
Molecular Weight (g/mol) 193.25
PubChem CID 137930
Formula Weight 131.18
Percent Purity 98%
Chemical Name or Material 6-Methylindole
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'