missing translation for 'onlineSavingsMsg'
Learn More

6-morpholin-4-ylpyridazine-3-carboxylic acid hydrochloride hydrate, Thermo Scientific™

Product Code. 10753717
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1192758-40-9
Molecular Formula C9H14ClN3O4
Molecular Weight (g/mol) 263.678
InChI Key VARLCSQEQQKXKH-UHFFFAOYSA-N
Synonym 6-morpholinopyridazine-3-carboxylic acid hydrochloride hydrate, 6-morpholin-4-ylpyridazine-3-carboxylic acid hydrochloride hydrate, 6-morpholin-4-yl pyridazine-3-carboxylic acid hydrate hydrochloride
PubChem CID 53444630
IUPAC Name 6-morpholin-4-ylpyridazine-3-carboxylic acid;hydrate;hydrochloride
SMILES C1COCCN1C2=NN=C(C=C2)C(=O)O.O.Cl

Specifications

Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 245.67
Percent Purity 97%
Chemical Name or Material 6-morpholin-4-ylpyridazine-3-carboxylic acid hydrochloride hydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'