missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Nitro-1,3-dihydroisobenzofuran-1-one, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
50 g
100 g
Specifications
Specifications
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 100 g |
Formula Weight | 179.13 |
Percent Purity | 97% |
Chemical Name or Material | 6-Nitro-1, 3-dihydroisobenzofuran-1-one |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'