missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Nitro-1,3-dihydroisobenzofuran-1-one, 97%, Thermo Scientific™
£57.55 - £338.00
Chemical Identifiers
CAS | 610-93-5 |
---|---|
Molecular Formula | C8H5NO4 |
Molecular Weight (g/mol) | 179.13 |
MDL Number | MFCD00033529 |
InChI Key | RNWGZXAHUPFXLL-UHFFFAOYSA-N |
Synonym | 6-nitrophthalide, 6-nitroisobenzofuran-1 3h-one, 6-nitro-3h-isobenzofuran-1-one, 6-nitro-1,3-dihydroisobenzofuran-1-one, 6-nitro-1 3h-isobenzofuranone, 6-nitro-2-benzofuran-1 3h-one, 1 3h-isobenzofuranone, 6-nitro, 6-nitro-3-hydroisobenzofuran-1-one, 6-nitro-1,3-dihydro-2-benzofuran-1-one, 6-nitrophtalide |
PubChem CID | 223584 |
IUPAC Name | 6-nitro-1,3-dihydro-2-benzofuran-1-one |
SMILES | [O-][N+](=O)C1=CC=C2COC(=O)C2=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10255864
|
Thermo Scientific Maybridge
KM08350EA |
10 g |
£57.55
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10799035
|
Thermo Scientific Maybridge
KM08350EE |
50 g |
£215.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10250405
|
Thermo Scientific Maybridge
KM08350FA |
100 g |
£338.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
610-93-5 | |
179.13 | |
RNWGZXAHUPFXLL-UHFFFAOYSA-N | |
223584 | |
[O-][N+](=O)C1=CC=C2COC(=O)C2=C1 |
C8H5NO4 | |
MFCD00033529 | |
6-nitrophthalide, 6-nitroisobenzofuran-1 3h-one, 6-nitro-3h-isobenzofuran-1-one, 6-nitro-1,3-dihydroisobenzofuran-1-one, 6-nitro-1 3h-isobenzofuranone, 6-nitro-2-benzofuran-1 3h-one, 1 3h-isobenzofuranone, 6-nitro, 6-nitro-3-hydroisobenzofuran-1-one, 6-nitro-1,3-dihydro-2-benzofuran-1-one, 6-nitrophtalide | |
6-nitro-1,3-dihydro-2-benzofuran-1-one |
Specifications
610-93-5 | |
Amber glass bottle | |
MFCD00033529 | |
RNWGZXAHUPFXLL-UHFFFAOYSA-N | |
6-nitro-1,3-dihydro-2-benzofuran-1-one | |
223584 | |
97% |
97% | |
C8H5NO4 | |
6-nitrophthalide, 6-nitroisobenzofuran-1 3h-one, 6-nitro-3h-isobenzofuran-1-one, 6-nitro-1,3-dihydroisobenzofuran-1-one, 6-nitro-1 3h-isobenzofuranone, 6-nitro-2-benzofuran-1 3h-one, 1 3h-isobenzofuranone, 6-nitro, 6-nitro-3-hydroisobenzofuran-1-one, 6-nitro-1,3-dihydro-2-benzofuran-1-one, 6-nitrophtalide | |
[O-][N+](=O)C1=CC=C2COC(=O)C2=C1 | |
179.13 | |
179.13 | |
6-Nitro-1, 3-dihydroisobenzofuran-1-one |