missing translation for 'onlineSavingsMsg'
Learn More

6-Nitroindole-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 15409478
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10242-00-9
Molecular Formula C9H6N2O4
Molecular Weight (g/mol) 206.157
MDL Number MFCD02664466
InChI Key JIXJNWVLWQQNDB-UHFFFAOYSA-N
Synonym 6-nitroindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 6-nitro, 6-nitro-2-indolecarboxylic acid, 1h-indole-2-carboxylicacid, 6-nitro, 6-nitroindol-2-carboxylic acid
PubChem CID 4714997
IUPAC Name 6-nitro-1H-indole-2-carboxylic acid
SMILES C1=CC2=C(C=C1[N+](=O)[O-])NC(=C2)C(=O)O

Specifications

Quantity 1 g
Formula Weight 206.15
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 6-Nitroindole-2-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'