missing translation for 'onlineSavingsMsg'
Learn More

6'-O-Apiosyl-5-O-Methylvisammioside, MedChemExpress

Product Code. p-200096275
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 16452589

Brand: Medchem Express HYN2295

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 6'-O-Apiosyl-5-O-Methylvisammioside is one of the components of the traditional Chinese medicine Kang-Jing.

6'-O-Apiosyl-5-O-Methylvisammioside is one of the components of the traditional Chinese medicine Kang-Jing.

Specifications

Chemical Name or Material 6'-O-Apiosyl-5-O-Methylvisammioside
CAS 139446-82-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C27H36O14
Quantity 1 mg
SMILES COC1=C(C[C@@H](C(C)(C)O[C@@H]([C@@H]([C@@H](O)[C@@H]2O)O)O[C@@H]2CO[C@H](OC[C@]3(O)CO)[C@@H]3O)O4)C4=CC(OC(C)=C5)=C1C5=O
Molecular Weight (g/mol) 584.57
Formula Weight 584.57
Percent Purity 99.87%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.