missing translation for 'onlineSavingsMsg'
Learn More

6-Phenoxynicotinaldehyde, 97%, Thermo Scientific™

£38.00 - £163.00

Chemical Identifiers

CAS 173282-69-4
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.209
MDL Number MFCD02681950
InChI Key SOPOQXNWJNVZMI-UHFFFAOYSA-N
PubChem CID 2776500
IUPAC Name 6-phenoxypyridine-3-carbaldehyde
SMILES C1=CC=C(C=C1)OC2=NC=C(C=C2)C=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10551512
View Documents
Thermo Scientific Maybridge
CC19604CB
250 mg Amber Glass Bottle
£38.00
250mg
Please to purchase this item. Need a web account? Register with us today!
10487032
View Documents
Thermo Scientific Maybridge
CC19604DA
1 g Amber Glass Bottle
£116.00
1g
Please to purchase this item. Need a web account? Register with us today!
10774755
View Documents
Thermo Scientific Maybridge
CC19604DE
5 g Amber Glass Bottle
£163.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

173282-69-4
199.209
SOPOQXNWJNVZMI-UHFFFAOYSA-N
6-phenoxypyridine-3-carbaldehyde
C12H9NO2
MFCD02681950
2776500
C1=CC=C(C=C1)OC2=NC=C(C=C2)C=O

Specifications

173282-69-4
100.0
C12H9NO2
6-phenoxynicotinaldehyde, 6-phenoxypyridine-3-carboxaldehyde, 2-phenoxypyridine-5-carboxaldehyde, 3-pyridinecarboxaldehyde,6-phenoxy, 5-formyl-2-phenoxypyridine, 6-phenoxy-3-pyridinecarboxaldehyde, 6-phenoxy-pyridine-3-carbaldehyde, 3-pyridinecarboxaldehyde, 6-phenoxy
C1=CC=C(C=C1)OC2=NC=C(C=C2)C=O
199.209
199.21
6-Phenoxynicotinaldehyde
96.5
97%
MFCD02681950
SOPOQXNWJNVZMI-UHFFFAOYSA-N
6-phenoxypyridine-3-carbaldehyde
2776500
97%
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.