missing translation for 'onlineSavingsMsg'
Learn More

6-Phenylnicotinic acid, 95%, Thermo Scientific™

Product Code. 10487222
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 29051-44-3
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.209
MDL Number MFCD03086128
InChI Key DLFLQXUYRFIFOK-UHFFFAOYSA-N
Synonym 6-phenylnicotinic acid, 3-pyridinecarboxylic acid, 6-phenyl, 6-phenyl-nicotinic acid, 2-phenyl pyridine-5-carboxylic acid, 6-phenyl nicotinic acid, acmc-1cmxf, 2-phenyl-5-carboxypyridine, 3-carboxy-6-phenylpyridine, 2-phenyl-5-pyridinecarboxylic acid, 6-phenyl-3-pyridinecarboxylic acid
PubChem CID 120118
IUPAC Name 6-phenylpyridine-3-carboxylic acid
SMILES C1=CC=C(C=C1)C2=NC=C(C=C2)C(=O)O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 199.21
Percent Purity 95%
Chemical Name or Material 6-Phenylnicotinic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'