Learn More
6-Phenylnicotinic acid, 95%, Thermo Scientific™
£48.35 - £140.00
Chemical Identifiers
CAS | 29051-44-3 |
---|---|
Molecular Formula | C12H9NO2 |
Molecular Weight (g/mol) | 199.209 |
MDL Number | MFCD03086128 |
InChI Key | DLFLQXUYRFIFOK-UHFFFAOYSA-N |
Synonym | 6-phenylnicotinic acid, 3-pyridinecarboxylic acid, 6-phenyl, 6-phenyl-nicotinic acid, 2-phenyl pyridine-5-carboxylic acid, 6-phenyl nicotinic acid, acmc-1cmxf, 2-phenyl-5-carboxypyridine, 3-carboxy-6-phenylpyridine, 2-phenyl-5-pyridinecarboxylic acid, 6-phenyl-3-pyridinecarboxylic acid |
PubChem CID | 120118 |
IUPAC Name | 6-phenylpyridine-3-carboxylic acid |
SMILES | C1=CC=C(C=C1)C2=NC=C(C=C2)C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10712473
|
Thermo Scientific Maybridge
CC21101CB |
250 mg | Amber Glass Bottle |
£48.35
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10487222
|
Thermo Scientific Maybridge
CC21101DA |
1 g | Amber Glass Bottle |
£140.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
29051-44-3 | |
199.209 | |
DLFLQXUYRFIFOK-UHFFFAOYSA-N | |
120118 | |
C1=CC=C(C=C1)C2=NC=C(C=C2)C(=O)O |
C12H9NO2 | |
MFCD03086128 | |
6-phenylnicotinic acid, 3-pyridinecarboxylic acid, 6-phenyl, 6-phenyl-nicotinic acid, 2-phenyl pyridine-5-carboxylic acid, 6-phenyl nicotinic acid, acmc-1cmxf, 2-phenyl-5-carboxypyridine, 3-carboxy-6-phenylpyridine, 2-phenyl-5-pyridinecarboxylic acid, 6-phenyl-3-pyridinecarboxylic acid | |
6-phenylpyridine-3-carboxylic acid |
Specifications
29051-44-3 | |
100.0 | |
C12H9NO2 | |
6-phenylnicotinic acid, 3-pyridinecarboxylic acid, 6-phenyl, 6-phenyl-nicotinic acid, 2-phenyl pyridine-5-carboxylic acid, 6-phenyl nicotinic acid, acmc-1cmxf, 2-phenyl-5-carboxypyridine, 3-carboxy-6-phenylpyridine, 2-phenyl-5-pyridinecarboxylic acid, 6-phenyl-3-pyridinecarboxylic acid | |
C1=CC=C(C=C1)C2=NC=C(C=C2)C(=O)O | |
199.209 | |
199.21 | |
6-Phenylnicotinic acid |
94.5 | |
95% | |
MFCD03086128 | |
DLFLQXUYRFIFOK-UHFFFAOYSA-N | |
6-phenylpyridine-3-carboxylic acid | |
120118 | |
95% |
Safety and Handling
GHS P Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning