missing translation for 'onlineSavingsMsg'
Learn More

SCHOTT AG Lighting and Imaging™ 6-Point Ring Light

The 6-Point Ring Light provides optimal lighting for contrast while reducing shadows. Mounts directly to objective by means of thumb screw. For use with cold light source KL300LED.

Brand:  SCHOTT AG Lighting and Imaging™ 172601

Product Code. 15773032

  • £455.00 / 1 unit
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 6-point Ringlight with eco-friendly Puravis fiber optics.
  • Fiber bundle protected with flexible metal PVC sheathing.
  • Housing made of rugged, black anodized aluminum.
  • Cost effective in comparison to annular ringlights.
  • Illumination with reduced shadows, but still sufficient shadows for improved contrasts.
Specifications

Specifications

Fiber Optic Ringlight
94 mm
Stereo Microscopy illumination
0.9 in.
39.4 in.
Eco-friendly Puravis Glass Optical Fibers
3.7 in.
66 mm
Cold light source KL300LED
23 mm
1000 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.