missing translation for 'onlineSavingsMsg'
Learn More

6-Quinoxalinamine, 97%, Thermo Scientific™

Product Code. p-2702855
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10354322

Brand: Thermo Scientific Maybridge CC11714DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 6298-37-9
Molecular Formula C8H7N3
Molecular Weight (g/mol) 145.17
MDL Number MFCD00462821
InChI Key MSGRFBKVMUKEGZ-UHFFFAOYSA-N
Synonym 6-aminoquinoxaline, 6-quinoxalinamine, 6-quinoxalinylamine, quinoxalin-6-ylamine, quinoxaline-6-ylamine, 6-chinoxalinamin, 6-amino-quinoxaline, pubchem15453, quinoxalin, 6-amino
PubChem CID 237859
IUPAC Name quinoxalin-6-amine
SMILES NC1=CC=C2N=CC=NC2=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 145.16
Percent Purity 97%
Chemical Name or Material 6-Quinoxalinamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.