missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ 6-ROX (6-Carboxy-X-Rhodamine), single isomer
6-ROX (6-Carboxy-X-Rhodamine), single isomer
Brand: Molecular Probes™ C6156
186.12 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
The excitation and emission spectra of carboxyrhodamine 6G (CR 6G) fall between those of fluorescein and tetramethylrhodamine. With a peak absorption at 525 nm, conjugates of carboxyrhodamine 6G are an excellent match to the 514 nm spectral line of the argon-ion laser. The amine-reactive 6-CR 6G, SE can be used to attach this fluorophore to biomolecules of interest including amine-modified oligonucleotides.Specifications
10 mg | |
Store in freezer (-5 to -30°C) and protect from light. | |
Room Temperature | |
ROX | |
525 |
6-ROX | |
Amine | |
Carboxylic Acid | |
Classic Dyes | |
514 |
For Research Use Only. Not for use in diagnostic procedures.