missing translation for 'onlineSavingsMsg'
Learn More
6-(tert-Butyl) 3-ethyl 2-amino-4,7-dihydrothieno[2,3-c]pyridine-3,6(5H)-dicarboxylate, Thermo Scientific™ Product Code.: 10433763

6-(tert-Butyl) 3-ethyl 2-amino-4,7-dihydrothieno[2,3-c]pyridine-3,6(5H)-dicarboxylate, Thermo Scientific™

Product Code. 10433763
1 g, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10433763

Brand: Thermo Scientific Maybridge MO01303DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 193537-14-3
Molecular Formula C15H22N2O4S
Molecular Weight (g/mol) 326.411
InChI Key XSXVOVXVHBSSSN-UHFFFAOYSA-N
Synonym ethyl 6-boc-2-amino-4,7-dihydro-5h-thieno 2,3-c pyridine-3-carboxylate, 2-amino-4,7-dihydro-5h-thieno 2,3-c pyridine-3,6-dicarboxylic acid 6-tert butyl ester 3-ethyl ester, 6-tert-butyl 3-ethyl 2-amino-4,5-dihydrothieno 2,3-c pyridine-3,6 7h-dicarboxylate, 6-tert-butyl 3-ethyl 2-amino-4h,5h,6h,7h-thieno 2,3-c pyridine-3,6-dicarboxylate, 6-tert-butyl 3-ethyl 2-amino-4,7-dihydrothieno 2,3-c pyridine-3,6 5h-dicarboxylate, 2-amino-6-boc-4,5,6,7-tetrahydro-thieno 2,3-c pyridine-3-carboxylic acid ethyl ester, 2-amino-4,7-dihydro-5h-thieno 2,3-c pyridine-3,6-dicarboxylic acid 6-tert-butyl ester 3-ethyl ester, 6-tert-butyl 3-ethyl 2-amino-4h,5h,7h-thieno 2,3-c pyridine-3,6-dicarboxylate, ethyl 2-amino-4,5,6,7-tetrahydrothieno 2,3-c pyridine-3-carboxylate, n-boc protectedShow MoreShow Less
PubChem CID 2794740
IUPAC Name 6-O-tert-butyl 3-O-ethyl 2-amino-5,7-dihydro-4H-thieno[2,3-c]pyridine-3,6-dicarboxylate
SMILES CCOC(=O)C1=C(SC2=C1CCN(C2)C(=O)OC(C)(C)C)N

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 326.41
Percent Purity 95%
Chemical Name or Material 6-(tert-Butyl) 3-ethyl 2-amino-4,7-dihydrothieno[2,3-c]pyridine-3,6(5H)-dicarboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6-(tert-Butyl) 3-ethyl 2-amino-4,7-dihydrothieno[2,3-c]pyridine-3,6(5H)-dicarboxylate, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.